imidazo[1,2-a]pyridine-6-carbonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Imidazo[1,2-a]pyridine-6-carbonitrile, 6-Cyanoimidazo[1,2-a]pyridine

OR4430-1G OR4430-250MG OR4430-5G
APOSOR4430-1GEA 27.7 EUR
APOSOR4430-1G APOSOR4430-250MG APOSOR4430-5G
imidazo[1,2-a]pyridine-6-carbonitrile 97%
imidazo[1,2-a]pyridine-6-carbonitrile
Formel: C₈H₅N₃
Molecular Weight: 143,149 g/mol
Schmelzpunkt: 160…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659603
CAS-Nummer: 106850-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR