3-Bromo-2-chloropyridine-5-sulphonyl chloride 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Bromo-6-chloropyridine-3-sulphonyl chloride

OR4475-1G
APOSOR4475-1GEA 74.1 EUR
APOSOR4475-1G
3-Bromo-2-chloropyridine-5-sulphonyl chloride 96%
3-Bromo-2-chloropyridine-5-sulphonyl chloride
Formel: C₅H₂BrCl₂NO₂S
Molecular Weight: 290.96 g/mol
Schmelzpunkt: 65…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318107
CAS-Nummer: 216394-05-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR