5-Brom-2-nitropyridin

Lieferant: Apollo Scientific
Achtung

OR4485-100G OR4485-25G OR4485-5G
APOSOR4485-100GEA 116 EUR
APOSOR4485-100G APOSOR4485-25G APOSOR4485-5G
5-Brom-2-nitropyridin
5-Brom-2-nitropyridin
Formel: C₅H₃BrN₂O₂
Molecular Weight: 203 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00160411
CAS-Nummer: 39856-50-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR