2-Hydroxy-4,6-dimethylnicotinic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-Dimethyl-2-hydroxynicotinic acid, 4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

OR4491-1G OR4491-100MG OR4491-5G
APOSOR4491-1GEA 137 EUR
APOSOR4491-1G APOSOR4491-100MG APOSOR4491-5G
2-Hydroxy-4,6-dimethylnicotinic acid 95%
2-Hydroxy-4,6-dimethylnicotinic acid
Formel: C₈H₉NO₃
Molecular Weight: 167,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234425
CAS-Nummer: 24667-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR