4-Hydroxy-2-methylpyridine 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methylpyridin-4-ol
OR450008-10G
OR450008-25G
OR450008-5G
APOSOR450008-10GEA
105
EUR
APOSOR450008-10G
APOSOR450008-25G
APOSOR450008-5G
4-Hydroxy-2-methylpyridine 98%
4-Hydroxy-2-methylpyridine
Formel:
C₆H₇NO Molecular Weight: 109.1259 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114156 CAS-Nummer: 18615-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...