Ethyl-4-methyl-1H-imidazol-5-carboxylat 95+%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-Methyl-1H-imidazole-5-carboxylate, 4-Methyl-1H-imidazole-5-carboxylic acid ethyl ester, Ethyl 4-methyl-5-imidazolecarboxylate, Ethyl 5-methyl-1H-imidazole-4-carboxylate
OR450016-100G
APOSOR450016-100GEA
191
EUR
APOSOR450016-100G
Ethyl-4-methyl-1H-imidazol-5-carboxylat 95+%
Ethyl-5-methyl-1H-imidazole-4-carboxylate
Formel:
C₇H₁₀N₂O₂ Molecular Weight: 154,1665 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005199 CAS-Nummer: 51605-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...