5-Bromoisoindoline hydrochloride 97%

Lieferant: Apollo Scientific

OR450024-1G OR450024-5G
APOSOR450024-1GEA 79.9 EUR
APOSOR450024-1G APOSOR450024-5G
5-Bromoisoindoline hydrochloride 97%
5-Bromoisoindoline hydrochloride
Formel: C₈H₉BrClN
Molecular Weight: 234,5200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10700135
CAS-Nummer: 919346-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR