3-Mercapto-5-oxo-4,5-dihydro-1,2,4-triazine-6-carboxylic acid
Lieferant: Apollo Scientific
OR450088-1G
OR450088-250MG
APOSOR450088-1GEA
377
EUR
APOSOR450088-1G
APOSOR450088-250MG
3-Mercapto-5-oxo-4,5-dihydro-1,2,4-triazine-6-carboxylic acid
3-Mercapto-5-oxo-4,5-dihydro-1,2,4-triazine-6-carboxylic acid
Formel:
C₄H₃N₃O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD01573074 CAS-Nummer: 7338-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...