1-Acetylazetidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Acetyl-3-azetidinecarboxylic Acid

OR451028-5G OR451028-1G
APOSOR451028-5GEA 342 EUR
APOSOR451028-5G APOSOR451028-1G
1-Acetylazetidine-3-carboxylic acid
1-Acetylazetidine-3-carboxylic acid
Formel: C₆H₉NO₃
Molecular Weight: 143,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406968
CAS-Nummer: 97628-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR