Methanesulphonylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Methylsulphonyl)acetic acid

OR45106-1G OR45106-25G OR45106-5G
APOSOR45106-1GEA 29 EUR
APOSOR45106-1G APOSOR45106-25G APOSOR45106-5G
Methanesulphonylacetic acid
Methanesulphonylacetic acid
Formel: C₃H₆O₄S
Molecular Weight: 138.14 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051843
CAS-Nummer: 2516-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR