5-Chinolincarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR451147-5G
OR451147-1G
OR451147-10G
APOSOR451147-5GEA
66.7
EUR
APOSOR451147-5G
APOSOR451147-1G
APOSOR451147-10G
5-Chinolincarbonsäure
5-Chinolincarbonsäure
Formel:
C₁₀H₇NO₂ Molecular Weight: 173,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047608 CAS-Nummer: 7250-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...