4-Spiroindenepiperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Spiro[indene-1,4'-piperidine] hydrochloride

OR451149-1G OR451149-5G
APOSOR451149-1GEA 430 EUR
APOSOR451149-1G APOSOR451149-5G
4-Spiroindenepiperidine hydrochloride
4-Spiroindenepiperidine hydrochloride
Formel: C₁₃H₁₆ClN
Molecular Weight: 221.73 g/mol
Schmelzpunkt: 291…294 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632538
CAS-Nummer: 137730-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR