4-Bromophenyl cyclobutyl ketone 98%

Lieferant: Apollo Scientific

Synonyme: (4-Bromophenyl)cyclobutyl-methanone

OR451175-5G OR451175-1G
APOSOR451175-5GEA 786 EUR
APOSOR451175-5G APOSOR451175-1G
4-Bromophenyl cyclobutyl ketone 98%
4-Bromophenyl cyclobutyl ketone
Formel: C₁₁H₁₁BrO
Molecular Weight: 239,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03841260
CAS-Nummer: 898790-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR