2-Brom-4-iodanisol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-iodo-1-methoxybenzene
OR45120-1G
OR45120-5G
APOSOR45120-1GEA
30.6
EUR
APOSOR45120-1G
APOSOR45120-5G
2-Brom-4-iodanisol
2-Brom-4-iodanisol
Formel:
C₇H₆BrIO Molecular Weight: 312,9304 g/mol Schmelzpunkt: 89…92 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03425935 CAS-Nummer: 182056-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...