(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-Cpg-OH·DCHA
OR451205-1G
OR451205-5G
APOSOR451205-1GEA
97.3
EUR
APOSOR451205-1G
APOSOR451205-5G
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid
Formel:
C₁₂H₂₁NO₄ Molecular Weight: 243,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663437 CAS-Nummer: 109183-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...