O-Benzyl-DL-serine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-OBzl-Ser-OH, H-DL-Ser(Bzl)-OH

OR45125-25G OR45125-5G
APOSOR45125-25GEA 249 EUR
APOSOR45125-25G APOSOR45125-5G
O-Benzyl-DL-serine 96%
O-Benzyl-DL-serine
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,22 g/mol
Schmelzpunkt: 225 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00021724
CAS-Nummer: 5445-44-3
UN: 1728
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR