(2-Methoxyphenoxy)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Methoxyphenoxy)acetic acid
OR451310-100G
OR451310-25G
OR451310-5G
APOSOR451310-100GEA
265
EUR
APOSOR451310-100G
APOSOR451310-25G
APOSOR451310-5G
(2-Methoxyphenoxy)essigsäure 98%
(2-Methoxyphenoxy)essigsäure
Formel:
(C₆H₄OCH₃)OCH₂COOH Molecular Weight: 182.18 g/mol Schmelzpunkt: 123…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014352 CAS-Nummer: 1878-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...