3,5-Dibromsalicylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dibromo-2-hydroxybenzoic acid

OR4514-100G OR4514-25G OR4514-5G
APOSOR4514-100GEA 80 EUR
APOSOR4514-100G APOSOR4514-25G APOSOR4514-5G
3,5-Dibromsalicylsäure
3,5-Dibromsalicylsäure
Formel: C₇H₄Br₂O₃
Molecular Weight: 295,91 g/mol
Schmelzpunkt: 223…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002441
CAS-Nummer: 3147-55-5
EINECS: 221-570-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR