(5-Methyl-1H-pyrazol-1-yl)essigsäure

Lieferant: Apollo Scientific
Achtung

OR451406-1G OR451406-5G
APOSOR451406-1GEA 33.5 EUR
APOSOR451406-1G APOSOR451406-5G
(5-Methyl-1H-pyrazol-1-yl)essigsäure
(5-Methyl-1H-pyrazol-1-yl)essigsäure
Formel: C₆H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03419599
CAS-Nummer: 180741-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR