1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Cyclopropyl-3-(dimethylamino)prop-2-en-1-one
OR451414-1G
OR451414-5G
APOSOR451414-1GEA
58
EUR
APOSOR451414-1G
APOSOR451414-5G
1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one
1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one
Formel:
C₈H₁₃NO Molecular Weight: 139,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840950 CAS-Nummer: 21666-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...