4-Amino-2-isopropyl-5-methylphenol hydrochloride

Lieferant: Apollo Scientific
Achtung

OR451415-1G OR451415-5G
APOSOR451415-1GEA 21.9 EUR
APOSOR451415-1G APOSOR451415-5G
4-Amino-2-isopropyl-5-methylphenol hydrochloride
4-Amino-2-isopropyl-5-methylphenol hydrochloride
Formel: C₁₀H₁₆ClNO
Molecular Weight: 201.7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035504
CAS-Nummer: 6321-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR