5-Bromoquinolin-8-ylamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR451420-1G
OR451420-5G
APOSOR451420-1GEA
31.9
EUR
APOSOR451420-1G
APOSOR451420-5G
5-Bromoquinolin-8-ylamine hydrochloride
5-Bromoquinolin-8-ylamine hydrochloride
Formel:
C₉H₈BrClN₂ Molecular Weight: 259.54 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00457010 CAS-Nummer: 297760-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...