1-Methyl-2-piperidin-1-ylethylamine

Lieferant: Apollo Scientific
Gefahr

OR451433-1G OR451433-5G
APOSOR451433-1GEA 348 EUR
APOSOR451433-1G APOSOR451433-5G
1-Methyl-2-piperidin-1-ylethylamine
1-Methyl-2-piperidin-1-ylethylamine
Formel: C₈H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01318922
CAS-Nummer: 34217-60-2
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR