4-Brom-1-ethyl-1H-pyrazol-3-carbaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR451435-1G
OR451435-25G
OR451435-5G
APOSOR451435-1GEA
23.2
EUR
APOSOR451435-1G
APOSOR451435-25G
APOSOR451435-5G
4-Brom-1-ethyl-1H-pyrazol-3-carbaldehyd
4-Brom-1-ethyl-1H-pyrazol-3-carbaldehyd
Formel:
C₆H₇BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419804 CAS-Nummer: 1001519-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...