4-Bromo-2-ethyl-2H-pyrazole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR451436-1G OR451436-25G OR451436-5G
APOSOR451436-1GEA 161 EUR
APOSOR451436-1G APOSOR451436-25G APOSOR451436-5G
4-Bromo-2-ethyl-2H-pyrazole-3-carbaldehyde
4-Bromo-2-ethyl-2H-pyrazole-3-carbaldehyde
Formel: C₆H₇BrN₂O
Molecular Weight: 203.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419806
CAS-Nummer: 1001519-37-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR