3-Cyclopropyl-1H-pyrazole-5-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR451446-1G
OR451446-5G
APOSOR451446-1GEA
65.4
EUR
APOSOR451446-1G
APOSOR451446-5G
3-Cyclopropyl-1H-pyrazole-5-carbaldehyde
3-Cyclopropyl-1H-pyrazole-5-carbaldehyde
Formel:
C₇H₈N₂O Molecular Weight: 136.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12548034 CAS-Nummer: 1284220-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...