Methyl-4-(chloromethyl)-5-methylisoxazole-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR451447-1G
OR451447-5G
OR451447-10G
APOSOR451447-1GEA
212
EUR
APOSOR451447-1G
APOSOR451447-5G
APOSOR451447-10G
Methyl-4-(chloromethyl)-5-methylisoxazole-3-carboxylate
Methyl-4-(chloromethyl)-5-methylisoxazole-3-carboxylate
Formel:
C₇H₈ClNO₃ Molecular Weight: 189,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14702898 CAS-Nummer: 1177292-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...