1-Ethylpyrrolidine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR451448-1G OR451448-5G
APOSOR451448-1GEA 197 EUR
APOSOR451448-1G APOSOR451448-5G
1-Ethylpyrrolidine-2-carboxylic acid
1-Ethylpyrrolidine-2-carboxylic acid
Formel: C₇H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11505336
CAS-Nummer: 165552-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR