(±)-3-Buten-2-ol

Lieferant: Apollo Scientific

Synonyme: (±)-Methylvinylcarbinol, (±)-1-Buten-3-ol, (±)-Methyl-vinylcarbinol, (±)-3-Hydroxy-1-buten

OR451801-25G
APOSOR451801-25GEA 89.9 EUR
APOSOR451801-25G
(±)-3-Buten-2-ol
(±)-3-Buten-2-ol
Formel: C₄H₈O
Molecular Weight: 72,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004543
CAS-Nummer: 598-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR