4-Piperidin-1-ylmethylbenzoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Piperidinylmethyl)-benzoic acid hydrochloride

OR452053-1G OR452053-5G
APOSOR452053-1GEA 307 EUR
APOSOR452053-1G APOSOR452053-5G
4-Piperidin-1-ylmethylbenzoic acid 97%
4-Piperidin-1-ylmethylbenzoic acid
Formel: C₁₃H₁₇NO₂
Molecular Weight: 219,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05240080
CAS-Nummer: 159691-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR