(3,4-Dichlorphenoxy)essigsäure 97%

Lieferant: Apollo Scientific

Synonyme: 3,4-D

OR452057-5G
APOSOR452057-5GEA 71.2 EUR
APOSOR452057-5G
(3,4-Dichlorphenoxy)essigsäure 97%
(3,4-Dichlorphenoxy)essigsäure
Formel: (C₆H₃Cl₂)OCH₂COOH
Molecular Weight: 221.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004303
CAS-Nummer: 588-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR