3-(4-Methylphenoxy)azetidine hydrochloride 97%

Lieferant: Apollo Scientific

OR452074-1G OR452074-250MG OR452074-500MG
APOSOR452074-1GEA 654 EUR
APOSOR452074-1G APOSOR452074-250MG APOSOR452074-500MG
3-(4-Methylphenoxy)azetidine hydrochloride 97%
3-(4-Methylphenoxy)azetidine hydrochloride
Formel: C₁₀H₁₄ClNO
Molecular Weight: 199,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16613952
CAS-Nummer: 1228070-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR