Pyrrolidine-3-carboxylic acid methylamide hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: N-Methylpyrrolidine-3-carboxamide hydrochloride, N-Methyl-3-pyrrolidinecarboxamide hydrochloride

OR452107-1G OR452107-250MG
APOSOR452107-1GEA 163 EUR
APOSOR452107-1G APOSOR452107-250MG
Pyrrolidine-3-carboxylic acid methylamide hydrochloride 97%
Pyrrolidine-3-carboxylic acid methylamide hydrochloride
Formel: C₆H₁₃ClN₂O
Molecular Weight: 164.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21605763
CAS-Nummer: 1361115-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR