1-Benzoylpiperidine-3-carboxylic acid 97%
Lieferant: Apollo Scientific
OR452127-1G
OR452127-500MG
OR452127-5G
APOSOR452127-1GEA
139
EUR
APOSOR452127-1G
APOSOR452127-500MG
APOSOR452127-5G
1-Benzoylpiperidine-3-carboxylic acid 97%
1-Benzoylpiperidine-3-carboxylic acid
Formel:
C₁₃H₁₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01856779 CAS-Nummer: 13850-76-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...