mono-Fmoc ethylene diamine hydrochloride 99%

Lieferant: Apollo Scientific

Synonyme: (9H-Fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrochloride, N-Fmoc-ethylenediamine hydrochloride, Fmoc-NH(CH2)2NH2·HCl

OR452155-5G
APOSOR452155-5GEA 109 EUR
APOSOR452155-5G
mono-Fmoc ethylene diamine hydrochloride 99%
N-FMOC-ethylenediamine hydrochloride
Formel: C₁₇H₁₈N₂O₂·HCl
Molecular Weight: 318,80 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00830741
CAS-Nummer: 391624-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR