tert-Butyl-2-hydroxy-2-phenylethylcarbamate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-2-hydroxy-2-phenylethylamine

OR452160-1G OR452160-500MG
APOSOR452160-1GEA 249 EUR
APOSOR452160-1G APOSOR452160-500MG
tert-Butyl-2-hydroxy-2-phenylethylcarbamate 97%
tert-Butyl-2-hydroxy-2-phenylethylcarbamate
Formel: C₁₃H₁₉NO₃
Molecular Weight: 237.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750569
CAS-Nummer: 67341-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR