3-(4-Methylphenoxy)piperidine 97%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Methylphenoxy)-piperidine hydrochloride

OR452206-1G OR452206-500MG
APOSOR452206-1GEA 568 EUR
APOSOR452206-1G APOSOR452206-500MG
3-(4-Methylphenoxy)piperidine 97%
3-(4-Methylphenoxy)piperidine
Formel: C₁₂H₁₇NO
Molecular Weight: 191,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061070
CAS-Nummer: 63843-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR