[(6-Chloropyridin-3-yl)methyl]methylamine dihydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-(methylaminomethyl)pyridine

OR452214-1G
APOSOR452214-1GEA 266 EUR
APOSOR452214-1G
[(6-Chloropyridin-3-yl)methyl]methylamine dihydrochloride 97%
[(6-Chloropyridin-3-yl)methyl]methylamine dihydrochloride
Formel: C₇H₁₁Cl₃N₂
Molecular Weight: 229,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442229
CAS-Nummer: 120739-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR