2-Amino-1-(1-pyrrolidinyl)-1-ethanone hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme: 2-Amino-1-(pyrrolidin-1-yl)ethanone hydrochloride
OR452219-1G OR452219-5G
APOSOR452219-1GEA 66.7 EUR
APOSOR452219-1G APOSOR452219-5G
2-Amino-1-(1-pyrrolidinyl)-1-ethanone hydrochloride 97%
2-Amino-1-(1-pyrrolidinyl)-1-ethanone hydrochloride
Formel: C₆H₁₃ClN₂O
Molecular Weight: 164,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366742
CAS-Nummer: 35855-14-2