1,4-Dibrom-2-ethylbenzol
Lieferant: Apollo Scientific
|
Achtung
|
OR4524-1G
OR4524-25G
OR4524-5G
APOSOR4524-1GEA
34.8
EUR
APOSOR4524-1G
APOSOR4524-25G
APOSOR4524-5G
1,4-Dibrom-2-ethylbenzol
1,4-Dibromo-2-ethylbenzene
Formel:
C₈H₈Br₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672998 CAS-Nummer: 289039-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...