3,5-Dibrombenzaldehyd
Lieferant: Apollo Scientific
|
Gefahr
|
OR4526-100G
OR4526-25G
OR4526-5G
APOSOR4526-100GEA
166
EUR
APOSOR4526-100G
APOSOR4526-25G
APOSOR4526-5G
3,5-Dibrombenzaldehyd
3,5-Dibromobenzaldehyde
Formel:
C₇H₄Br₂O Schmelzpunkt: 89,7…91.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156887 CAS-Nummer: 56990-02-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...