3-(Methylsulfonyl)anilin Hydrochlorid 95+%

Lieferant: Apollo Scientific
Achtung

OR4558-1G OR4558-5G
APOSOR4558-1GEA 50.9 EUR
APOSOR4558-1G APOSOR4558-5G
3-(Methylsulfonyl)anilin Hydrochlorid 95+%
3-(Methylsulfonyl)anilin Hydrochlorid
Formel: C₇H₁₀ClNO₂S
Schmelzpunkt: 228…230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00216487
CAS-Nummer: 80213-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR