2-Methyl-2-propanyl-3-oxo-1-piperazincarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine
OR4569-1G
OR4569-5G
APOSOR4569-1GEA
21.9
EUR
APOSOR4569-1G
APOSOR4569-5G
2-Methyl-2-propanyl-3-oxo-1-piperazincarboxylat
1-Boc-3-oxopiperazine
Formel:
C₉H₁₆N₂O₃ Molecular Weight: 200,238 g/mol Schmelzpunkt: 156…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181069 CAS-Nummer: 76003-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...