(S)-1-Boc-3-hydroxypiperidin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected

OR4581-25G OR4581-5G OR4581-100G
APOSOR4581-25GEA 33.5 EUR
APOSOR4581-25G APOSOR4581-5G APOSOR4581-100G
(S)-1-Boc-3-hydroxypiperidin 98%
(S)-1-Boc-3-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201.27 g/mol
Schmelzpunkt: 34…40 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115307
CAS-Nummer: 143900-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR