3,5-Dimethylbenzolsulfonylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
m-Xylol-5-sulfonsäurechlorid, m-Xylol-5-sulfonylchlorid, 3,5-Dimethylbenzol-1-sulfonylchlorid, 3,5-Dimethylbenzolsulfonsäurechlorid, 3,5-Dimethylbenzolsulfochlorid
OR4582-1G
OR4582-25G
OR4582-5G
APOSOR4582-1GEA
26.1
EUR
APOSOR4582-1G
APOSOR4582-25G
APOSOR4582-5G
3,5-Dimethylbenzolsulfonylchlorid 97%
3,5-Dimethylbenzolsulfonylchlorid
Formel:
C₈H₉ClO₂S Molecular Weight: 204,68 g/mol Schmelzpunkt: 90…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094655 CAS-Nummer: 2905-27-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...