1-Benzyloxy-4-brombenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene
OR4586-100G
OR4586-250G
OR4586-500G
APOSOR4586-100GEA
116
EUR
APOSOR4586-100G
APOSOR4586-250G
APOSOR4586-500G
1-Benzyloxy-4-brombenzol 98%
1-Benzyloxy-4-brombenzol
Formel:
C₁₃H₁₁BrO Molecular Weight: 263.13 Siedepunkt: 166…167 °C/4mmHg Schmelzpunkt: 60…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00028016 CAS-Nummer: 6793-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...