5-Chloruracil

Lieferant: Apollo Scientific

Synonyme: 5-Chlor-2,4-dihydroxypyrimidin, Chloruracil

OR460039-100G
APOSOR460039-100GEA 68.3 EUR
APOSOR460039-100G
5-Chloruracil
5-Chloruracil
Formel: C₄H₃ClN₂O₂
Molecular Weight: 146,53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006019
CAS-Nummer: 1820-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR