Ethyl-(2-amino-1,3-thiazol-4-yl)(oxo)acetat

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Amino-4-thiazolyl)-2-oxoacetic acid ethyl ester, Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate, Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate

OR46025-100G OR46025-25G OR46025-5G
APOSOR46025-100GEA 91.6 EUR
APOSOR46025-100G APOSOR46025-25G APOSOR46025-5G
Ethyl-(2-amino-1,3-thiazol-4-yl)(oxo)acetat
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate
Formel: C₇H₉NO₄S
Molecular Weight: 200.2150 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00010414
CAS-Nummer: 64987-08-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR