3,5-Dichloro-4-pyridinecarboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloro-4-formylpyridine, 3,5-Dichloropyridine-4-carboxaldehyde, 3,5-Dichloroisonicotinaldehyde

OR46033-1G OR46033-25G OR46033-5G
APOSOR46033-1GEA 21.9 EUR
APOSOR46033-1G APOSOR46033-25G APOSOR46033-5G
3,5-Dichloro-4-pyridinecarboxaldehyde
3,5-Dichloro-4-pyridinecarboxaldehyde
Formel: C₆H₃Cl₂NO
Molecular Weight: 176,00 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039921
CAS-Nummer: 136590-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR