2-Brom-6-chlorbenzaldehyd 99%

Lieferant: Apollo Scientific
Achtung

OR46035-100G OR46035-25G OR46035-5G
APOSOR46035-100GEA 427 EUR
APOSOR46035-100G APOSOR46035-25G APOSOR46035-5G
2-Brom-6-chlorbenzaldehyd 99%
2-Bromo-6-chlorobenzaldehyde
Formel: C₇H₄BrClO
Molecular Weight: 219,46 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00266792
CAS-Nummer: 64622-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR